Learn More
Trajan Scientific and Medical™ Pre-fitted Syringes with Valves
Product Code. 10285222
Click to view available options
Volume (Metric):
100 μL
Unit Size:
Each
Product Code. 10285222
Brand: Trajan Scientific and Medical™ 005279
For sample storage and transportation or to pressurize sample prior to injection
  • Accurate and reproducible injections
  • Pre-fitted with a valve for sample pressurization prior to injection
Specifications
Description Pre-fitted Syringe
Diameter (Metric) Outer Needle 0.63 mm
Includes Valve, Removable Needle
Volume (Metric) 100 μL