Learn More
Trajan Scientific and Medical™ Pre-fitted Syringes with Valves
Click to view available options
Volume (Metric):
100 μL
Unit Size:
Each
Description
- Accurate and reproducible injections
- Pre-fitted with a valve for sample pressurization prior to injection
Specifications
Specifications
| Description | Pre-fitted Syringe |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Includes | Valve, Removable Needle |
| Volume (Metric) | 100 μL |