missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PREB Recombinant Rabbit Monoclonal Antibody (JE64-56)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA544683

Product Code. 17827662

  • € 502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PREB Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Prolactin (PRL) expression in the pituitary is limited to specific cells. Pit-1 is a pituitary specific transcription factor that plays an important role in PRL expression, both in mature organism and during development. The PRL promoter contains numerous Pit-1 binding sites and these sites have been implicated in both basal level and kinase-mediated gene expression. The most proximal of these binding sites, termed 1P, has been shown to direct a response to numerous signals, such as cAMP and various G-proteins. A novel protein, termed PREB (prolactin regulatory element binding protein), has been recently identified that regulates PRL gene expression through the 1P site, though it contains no discernable DNA-binding motif. Recent studies suggest that PREB is encoded by a single-copy gene in both mice and humans and exhibits nuclear accumulation in pituitary cells. Evidence suggests that PREB is a novel transcription factor that assists in PRL expression whether alone, or in concert with Pit-1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PREB
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q9HCU5
Preb
Recombinant protein within human PREB aa 81-230/417.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
JE64-56
Unconjugated
Preb
C85705; mammalian guanine nucleotide exchange factor mSec12; PREB; prolactin regulatory binding-element protein; prolactin regulatory element binding; prolactin regulatory element binding protein; prolactin regulatory element-binding protein; Sec12
Rabbit
Protein A
RUO
10113
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.