Learn More
Schneider Ludwig™ Precision Density Hydrometer
Product Code. 10368274
Encompass_Preferred
Click to view available options
Length (Metric):
160 mm
250 mm
280 mm
300 mm
Specific Gravity Range:
0.800 to 0.900 g/cm3
0.880 to 0.940 g/cm3
0.900 to 1.000 g/cm3
0.940 to 1.000 g/cm3
1.000 to 1.100 g/cm3
1.000 to 1.200 g/cm3
1.000 to 1.500 g/cm3
1.100 to 1.200 g/cm3
1.200 to 1.300 g/cm3
Unit Size:
Each
Product Code. 10368274
Brand: Schneider Ludwig™ 3031007
Only null left
Specifications
Quantity 1Each
Type Precision Density Hydrometer
Length (Metric) 300 mm
Specific Gravity Range 1.200 to 1.300 g/cm3