missing translation for 'onlineSavingsMsg'
Learn More

Schneider Ludwig™ Precision Density Hydrometer

Brand:  Schneider Ludwig™ 3071003

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10647735

  • € 17.68 / Each
Estimated Shipment: 14-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

1Each
Precision Density Hydrometer
280 mm
1.000 to 1.200 g/cm3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers