Learn More
Bochem™ Precision Forceps
Manufactured from 18/10 stainless steel
€ 11.05 - € 90.75
Specifications
Tip Style | Sharp |
---|---|
No. per Pack | 1 Pack |
Material | Stainless Steel |
Length (Metric) | 120 mm |
Product Type | Forceps, Precision |
Product Code | Brand | Length (Metric) | Tip Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Tip Style | Price | Quantity & Availability | ||||
11895151
|
Bochem™
1640 |
120 mm | Sharp |
€ 90.75
Pack of 10 |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
10471601
|
Bochem™
1640 |
120 mm | Sharp |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Points without ridges
- Self-tensing
Specifications
Sharp | |
Stainless Steel | |
Forceps, Precision |
1 Pack | |
120 mm | |
Forceps |