missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Precision M1 Weight Set
Click to view available options
:
Precision M1 Weight Set
Description
- Precision calibration weight
Specifications
Specifications
| Product Type | Weight |
| Classification | M1 |
| Weight (Metric) | 1 to 500 mg |
| For Use With (Application) | Balance calibration |
| Quantity | 12 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction