Learn More
Ludwig Schneider Precision thermometer, ASTM 99 C - ACCU-SAFE -, round, -50+5°C, L: 35 mm,L: 320 mm, suit. for cal.
Brand:  Ludwig Schneider 1202099S
Encompass_Preferred
Precision thermometer, ASTM 99 C - ACCU-SAFE -, round, -50+5°C, L: 35 mm,L: 320 mm, suit. for cal.
Product Code. 16620606
  • € 204.97 / Pack of 1
Only null left
Estimated Shipment: 16-06-2026