Learn More
Ludwig Schneider Precision UK hydrometer L 50 SP , cal. at 15°C, 1.000-1.050 in 0.0005 gr/cm3, DAkkS-cal. cert. 3 p.
Brand:  Ludwig Schneider 3071135/04/3P
Encompass_Preferred
Precision UK hydrometer L 50 SP , cal. at 15°C, 1.000-1.050 in 0.0005 gr/cm3, DAkkS-cal. cert. 3 p.
Product Code. 16670676
  • € 360.85 / Each
Only null left
Estimated Shipment: 27-02-2026