missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pref-1/DLK1/FA1 Antibody (211309) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1144H
This item is not returnable.
View return policy
Description
Pref-1/DLK1/FA1 Monoclonal antibody specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Pref-1/DLK1/FA1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 211309 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| Mouse myeloma cell line NS0-derived recombinant human Pref-1 long isoform, Ala24-Pro297 (with Arg248Pro and Lys295Ser substitutions), Accession # P80370 | |
| 0.1 mL | |
| Signal Transduction | |
| 8788 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction