missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pref-1/DLK1/FA1 Antibody (MM0514-9D8), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11855PCP
This item is not returnable.
View return policy
Description
Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Pref-1/DLK1/FA1 | |
| Monoclonal | |
| PerCP | |
| P80370 | |
| DLK1 | |
| Human recombinant Pref-1/DLK1/FA1 | |
| 0.1 mL | |
| Signal Transduction | |
| 8788 | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry | |
| MM0514-9D8 | |
| Flow Cytometry | |
| delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction