Learn More
Honeywell™ Prefilter Pads
Prefilter Pads are used in conjunction with a retaining clip or adapter on Honeywell™ Twin-Filter Air-Purifying Respirators
Brand:  Honeywell™ 1002800
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11847171
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Product Type: Prefilter Pads
  • For Use With: Honeywell™ Twin-Filter Air-Purifying Respirators
Specifications
Specifications
Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators
EN 143
Requires Retaining Cover (1001590 or 1001606)
Prefilter Pad
Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators
20/Pk.
PPE Cat. 3; ISO 9001/2000
White
Filters T series
Respiratory Protection (P1, Dust)
TP1S