Learn More
Uvex Preformed Mask silv-Air c 2312 FFP3
Product Code. 17532965 Shop All Uvex Products
Click to view available options
:
Filter Type: FFP3
Product Code. 17532965
Brand: Uvex 8732312
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The silv-Air c product family consists of classic folding and cup-shaped face masks which fit different face shapes. Headband lacing system and integrated sealing lips in all protection classes make it easy to fit these masks correctly – and to wear them comfortably for long periods.
General Features
  • Particle-filtering preformed mask with exhalation valve – ergonomically adapted for smaller faces
  • Color: white
Protection Features
  • Certified FFP3 face mask to EN 149:2001+A1:2009
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Cannot be reused
Comfort Features
  • Four-point headband attachment and flexible, adjustable nose clip for a comfortable, secure fit
  • Very soft, all-around sealing lip for optimal wearer comfort and secure fit
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Ergonomically adjusted especially for smaller to medium faces
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Design Type Cup/Molded
Disposable Single-use
Material Polypropylene, Polyester
Size Small/Medium
With Exhalation Valve Yes
Filter Efficiency FFP3