Learn More
Uvex Preformed Mask silv-Air e 7233 FFP2
Shop All Uvex Products
Click to view available options
:
Filter Type: FFP2
Description
General Features
- Particle-filtering preformed mask with exhalation valve and inhalation valves
- Color: white
Protection Features
- Certified FFP2 face mask to EN 149:2001+A1:2009
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- R label (reusable): tested for re-usability and performance, including in the second shift
Comfort Features
- High-performance climazone system with three compartments for a comfortable, cool climate inside the mask
- Flexible textile headband with neck clip for individual adjustment
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
- Two extra filters with inhalation valves for lower breathing resistance and increased comfort due to easier air exchange
- Very soft, all-around sealing lip for optimum wearer comfort and secure fit
- Combination of 3D nose area and sealing lip ensures ideal fit for almost any nose shape
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 149 (R marked, D marked) |
| Product Type | Preformed Mask |
| With Exhalation Valve | Yes |
| Filter Efficiency | FFP2 |
| Includes | 360° exhalation valve, 3D nose section, circumferential comfort sealing lip |