Learn More
Uvex Preformed Mask silv-Air e 7317 FFP3, Individually Packed
Product Code. 17552955 Shop All Uvex Products
Change view
Click to view available options
:
Filter Type: FFP3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17552955 Filter Type: FFP3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17552955 Supplier Uvex Supplier No. 8707318
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All masks of the silv-Air e family combine the unique technologies of optimum wearer comfort with extremely low breathing resistance. The products are therefore suitable for heavy work and for being worn for long periods of time.
General Features
  • Particle-filtering preformed mask with exhalation valve
  • Color: white
Protective Features
  • FFP3 respirator certified in accordance with EN 149:2001+A1:2009
  • D symbol (mask has passed the dolomite dust test): breathe easy even at high levels of dust exposure
  • R certification (re-usable): tested for re-usability and performance during second shift
Comfort Features
  • Soft material edges for increased comfort
  • Very soft, all-around sealing lip for optimal wearer comfort and secure fit
  • No nose clip (metal-free)
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Extra-wide textile headband for enhanced comfort
  • Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (R marked, D marked)
Product Type Preformed Mask
With Exhalation Valve Yes
Filter Efficiency FFP3
Includes 360° exhalation valve, four-point head-securing strap, circumferential comfort sealing lip