Learn More
Uvex Preformed Mask silv-Air p 7110 FFP1
Shop All Uvex Products
Click to view available options
:
Filter Type: FFP1
Description
General Features
- Particle-filtering preformed mask with exhalation valve to reduce exhalation resistance and heat and moisture build-up inside the mask
- Metal-free processing
- Soft material edges for increased wearer comfort
- Seamless headband for a comfortable, secure fit
- Color: silver-gray
Protection Features
- FFP1 respirator certified in accordance with EN 149:2001+A1:2009
- D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
- Not reusable
Comfort Features
- Soft material edges for increased wearer comfort
- Seamless headband for a comfortable, secure fit
- Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 149 (D marked) |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Material | Polypropylene, Polyester |
| Product Type | Preformed Mask |
| Size | Standard |
| With Exhalation Valve | Yes |
| Filter Efficiency | FFP1 |
| Sterility | Non-sterile |