missing translation for 'onlineSavingsMsg'
Learn More

PRELID1 Monoclonal Antibody (OTI4A4), TrueMAB™, OriGene

Product Code. 17594544
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17594544 100 μL 100µL
17514554 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17594544 Supplier Origene Technologies Supplier No. TA810979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

May be important for the development of vital and immunocompetent organs.

Specifications

Antigen PRELID1
Applications Western Blot
Classification Monoclonal
Clone OTI4A4
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRELID1
Gene Accession No. Q9Y255
Gene Alias CGI-106, PRELI, PX19, SBBI12
Gene Symbols PRELID1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 36-219 of human PRELID1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27166
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.