Learn More
PRELID2 Monoclonal Antibody (OTI8F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810806
Encompass_Preferred
 View more versions of this product
Product Code. 17574294
  • € 540.87 / 100µL
Description
PRELID2 (PRELI Domain Containing 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatidic acid transporter activity. An important paralog of this gene is PRELID3B.
Specifications
Specifications
PRELID2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8N945
PRELID2
Full length human recombit protein of human PRELID2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI8F1
Unconjugated
PRELID2
PRELI domain containing 2
Mouse
Affinity Chromatography
RUO
153768
-20° C, Avoid Freeze/Thaw Cycles