missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRELP Antibody (01), FITC , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06179F
This item is not returnable.
View return policy
Description
PRELP Monoclonal antibody specifically detects PRELP in Human samples. It is validated for ELISA
Specifications
| PRELP | |
| Monoclonal | |
| FITC | |
| 55 kDa leucine-rich repeat protein of articular cartilage, MST161, MSTP161, prolargin, prolargin proteoglycan, proline arginine-rich end leucine-rich repeat protein, proline/arginine-rich end leucine-rich repeat protein, Proline-arginine-rich end leucine-rich repeat protein, SLRR2AMGC45323 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PRELP (NP_002716.1; Gln21-Ile382). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 01 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5549 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction