Learn More
Ark Plas Products™ Premium Grade Natural Kynar Barbed Threaded Connectors
Threaded barbed fittings for forming connections in the laboratory, with a pressure rating of up to 125 P.S.I. without clamps.
Brand:  Ark Plas Products™ MEX26-DF0
Additional Details : Weight : 0.01440kg
 View more versions of this product
Product Code. 11801160
  • € 24.92 / Pack of 10
Estimated Shipment
06-07-2021
Add to basket
Description
Description
  • Molded from virgin, lot-traceable material only
  • Manufactured to the most exacting standards
  • Made with critical use applications in mind
Specifications
Specifications
Natural
Barbed Threaded Connector
1/8 in. Barb
Kynar
10 Pack
1/8-27 MNPT