missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PREP Polyclonal Antibody

PREP Polyclonal Antibody
Brand: Thermo Scientific PA556723
This item is not returnable.
View return policy
Description
PREP Polyclonal Antibody for IHC
Specifications
| PREP | |
| Polyclonal | |
| Unconjugated | |
| PREP | |
| AI047692; AI450383; D10Wsu136e; dJ355L5.1 (prolyl endopeptidase); EC 3.4.21.26; MGC16060; PE; PE {ECO:0000250; PEP; Pop; post-proline cleaving enzyme; post-proline cleaving enzyme {ECO:0000250; Prep; prep {ECO:0000312; Prolyl endopeptidase; prolyl endopeptidase {ECO:0000250; prolyl oligopeptidase; RGD:620841}; rPop; UniProtKB:P23687} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5550 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48147 | |
| PREP | |
| Recombinant protein corresponding to Human PREP. Recombinant protein control fragment (Product #RP-94646). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction