Learn More
Invitrogen™ PREPL Recombinant Rabbit Monoclonal Antibody (24GB990)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557638
 View more versions of this product
Product Code. 30274853
  • € 564.00 / 100µL
Description
PREPL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PREPL
Recombinant Monoclonal
Unconjugated
Prepl
2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143
Rabbit
Affinity chromatography
RUO
213760, 298771, 9581
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB990
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q4J6C6, Q5HZA6, Q8C167
Prepl
A synthesized peptide derived from human PREPL (aa 50-100).