missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRICKLE2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 462.00
Specifications
| Antigen | PRICKLE2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PRICKLE2 Polyclonal antibody specifically detects PRICKLE2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PRICKLE2 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Stem Cells | |
| PBS with 50% glycerol, pH7.3. | |
| 166336 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp686D143, EPM5, prickle homolog 2 (Drosophila), Prickle Planar Cell Polarity Protein 2, prickle-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 615-844 of human PRICKLE2 (NP_942559.1). QEMEGNLHQLSNPIGYRDLQSHGRMHQSFDFDGGMAGSKLPGQEGVRIQPMSERTRRRATSRDDNRRFRPHRSRRSRRSRSDNALHLASEREAISRLKDRPPLRAREDYDQFMRQRSFQESMGHGSRRDLYGQCPRTVSDLALQNAFGDRWGPYFAEYDWCSTCSSSSESDNEGYFLGEPIPQPARLRYVTSDELLHKYSSYGLPKSSTLGGRGQLHSRKRQKSKNCIIS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title