missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRICKLE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545653
This item is not returnable.
View return policy
Description
Peptide sequence: IPQPARLRYV TSDELLHKYS SYGLPKSSTL GGRGQLHSRK RQKSKNCIIS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
This gene encodes a homolog of Drosophila prickle. The exact function of this gene is not known, however, studies in mice suggest that it may be involved in seizure prevention. Mutations in this gene are associated with progressive myoclonic epilepsy type 5.
Specifications
| PRICKLE2 | |
| Polyclonal | |
| Unconjugated | |
| PRICKLE2 | |
| 6230400G14Rik; 6720451F06Rik; DKFZp686D143; DKFZp686H1748; DKFZp686M031; EPM5; EPM5p; mpk2; PLP2; PRIC2; prickle homolog 2; prickle homolog 2 (Drosophila); prickle planar cell polarity protein 2; PRICKLE2; prickle-like 2; prickle-like protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 166336 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z3G6 | |
| PRICKLE2 | |
| Synthetic peptide directed towards the C-terminal of human PRICKLE2 (aa 795-844). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction