missing translation for 'onlineSavingsMsg'
Learn More

Pridopidine, MedChemExpress

Product Code. 16490116
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490116

Brand: Medchem Express HY10684

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pridopidine, a dopamine (DA) stabilizer, acts as a low affinity dopamine D2 receptor (D2R) antagonist. Pridopidine exerts high affinity towards sigma 1 receptor (S1R) with Ki between 70 and 80 nM, which is ∼100x higher than its affinity toward D2R.

Pridopidine, a dopamine (DA) stabilizer, acts as a low affinity dopamine D2 receptor (D2R) antagonist. Pridopidine exerts high affinity towards sigma 1 receptor (S1R) with Ki between 70 and 80 nM, which is ∼100x higher than its affinity toward D2R.

Specifications

Chemical Name or Material Pridopidine
CAS 346688-38-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H23NO2S
Quantity 2 mg
Synonym ACR16 ASP2314 FR310826
Solubility Information DMSO : 100 mg/mL (355.35 mM; Need ultrasonic)
SMILES O=S(C1=CC(C2CCN(CCC)CC2)=CC=C1)(C)=O
Molecular Weight (g/mol) 281.41
Formula Weight 281.41
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.