missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pridopidine
GREENER_CHOICE

Product Code. 18799946
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18799946

Brand: Tocris Bioscience™ 7630/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly selective σ1 agonist

Pridopidine is a selective high affinity σ-1 receptor (S1R) agonist (Ki = 7.1 nM in both HEK293 cells and rat striatal membranes). Pridopidine displays ∼30-fold and ∼100-fold selectivity over the dopamine D3 receptor and the D2 receptor, respectively. Pridopidine is a low affinity dopamine D2 receptor antagonist (Ki (low) = 17.5 μM; Ki (high) = 7.5 μM). Pridopidine upregulates gene expression induced by BDNF in rat striatum in vivo in an S1R-dependent manner. Pridopidine improves motor function and shows neuroprotective effects in Huntington's disease animal models. Pridopidine restores neuromuscular junction activity in amyotrophic lateral sclerosis models in vitro and in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[3-(Methylsulfonyl)phenyl]-1-propylpiperidine
CAS 346688-38-8
Quantity 10 mg
Target Sigma1 Receptor Agonists
Molecular Formula C15H23NO2S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.