missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRIM2 Monoclonal Antibody (4G2)
GREENER_CHOICE

Product Code. 15652215
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15652215 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15652215 Supplier Invitrogen™ Supplier No. MA524387

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MEFSGRKWRK LRLAGDQRNA SYPHCLQFYL QPPSENISLI EFENLAIDRV KLLKSVENLG VSYVKGTEQY QSKLESELRK LKFSYRENLE DEYEPRRRDH ISHFILRLAY CQSEELRRWF IQQEMDLLRF RFSILPKDKI QDFLKDSQLQ FEAVSIFL.

DNA primase is the polymerase that synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRIM2
Applications ELISA
Classification Monoclonal
Clone 4G2
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Prim2
Gene Accession No. P49643
Gene Alias AI323589; DNA polymerase alpha subunit III (primase); DNA primase 58 kDa subunit; DNA primase large subunit; DNA primase, p58 subunit; p58; Pola3; PRIM2; PRIM2A; primase (DNA) subunit 2; primase, DNA, polypeptide 2; primase, DNA, polypeptide 2 (58kDa)
Gene Symbols Prim2
Host Species Mouse
Immunogen PRIM2A (AAH17833, 1 a.a. ~ 158 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5558
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.