Learn More
Invitrogen™ PRIMER 5' DY549P1 200 NMOL
Product Code. 15490524
Click to view available options
:
200 nmol
Product Code. 15490524
Brand: Invitrogen™ 100032031
PRIMER 5' DY549P1 200 NMOL
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Description 5' DY54991 Primer, 200 nmol
Product Type 5' DY54991 Primer
Primer 5' Labeled
Quantity 200 nmol