Learn More
Invitrogen™ PRIMER 5' DY549P1 200 NMOL
Click to view available options
:
200 nmol
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
| Description | 5' DY54991 Primer, 200 nmol |
| Product Type | 5' DY54991 Primer |
| Primer | 5' Labeled |
| Quantity | 200 nmol |