Learn More
Carl Zeiss™ Primovert Microscope Stand with Binocular Tube
Description
- Primovert microscope stand with binocular tube
- 4-position objective nosepiece H
- Specimen stage 200x239 mm with stage insert
- Metal binocular tube 45°/20
- Eyepieces 10x/20 Br. foc.
- Transmitted-light illumination with 6V 30W halogen bulb
- Pinhole diaphragm and allen keys for adjusting phase contrast
- External power unit 100 to 240VAC/50 to 60Hz with country-specific adapters
- Includes dust cover
- 6V 30W halogen lamp (spare part)
Specifications
Specifications
| Type | Binocular Tube |
| Quantity | 1 Ea. |