Description
- Kit: Thermal printer (SRV000-0485) and 2 paper rolls 57 mm (THE002-0118).
- Used for cycle documentation.
- During the cycle the printer records temperatures, date, cycle status, end of cycle.
Specifications
Specifications
| Product Type | Printer |