Learn More
Interscience™ Printer Sticker Set
Click to view available options
Color:
White
Unit Size:
Pack of 2
Description
- Label size: 70 x 45 mm
Specifications
Specifications
| Product Type | Label |
| Color | White |
| For Use With (Equipment) | DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump Pro |
| Material | Thermal Printing Paper |
| Print Technology | Thermal Transfer |
| Autoclavable | Not Autoclavable |
| Quantity | 2 Rolls/Pk. |