Learn More
Epredia™ PrintMate™ Biopsy Cassettes
Shop All Epredia Products
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Conditionnement:
Pack of 1000
Pack of 750
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
Description
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.
Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
Spécification
Spécification
| Pore Style | Small Square Pore |
| PrintMate Compatible | Yes |
| Color | Pink |
| Product Type | Biopsy Cassette |