Learn More
Epredia™ PrintMate™ Slotted Cassettes in Pre-packed Tubes, Aqua
Product Code. 15477584 Shop All Epredia Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
missing translation for 'unitSize'
Pack of 1500
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15477584 Aqua Pack of 1500
15487584 Blue Pack of 1500
15497584 Green Pack of 1500
15407594 Gray Pack of 1500
15417594 Lilac Pack of 1500
15427594 Orange Pack of 1500
15447594 Pink Pack of 1500
15457594 Tan Pack of 1500
15467594 White Pack of 1500
15477594 Yellow Pack of 1500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 missing translation for 'options'
Product Code. 15477584 missing translation for 'mfr' Epredia™ missing translation for 'supplierNo' A84210030
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. Packed in tubes, PrintMate Cassettes are intended for high throughput printing, and provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slotted Pore
PrintMate Compatible Yes
Color Aqua
Product Type Biopsy Cassette