Learn More
PRKACA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27398-1-AP-20UL
This item is not returnable.
View return policy
Description
Protein kinase A (PKA, cAMP-dependent protein kinase) is a key element of a ubiquitous signaling pathway important in the cell cycle, cellular communication, memory formation and behavior. PKA is composed of two catalytic (PKAc; Protein Kinase A catalytic subunit) and two regulatory subunits (PKAr). Upon binding cAMP, the complex dissociates to PKAr dimer and two activated PKAc ser/thr protein kinase catalytic monomers. The released PKAc can translocate into the nucleus and exert a regulatory role in the activation of multiple nuclear hormone receptors. However, PKAc-mediated activation of tonicity-dependent gene expression is cAMP independent. Humans express three types of PKAc subunit - PKAc alpha is present in most human tissues, PKAc beta and gamma are tissue-specific, the later is found in testes.Specifications
| PRKACA | |
| Polyclonal | |
| Unconjugated | |
| PRKACA | |
| PKA C alpha, PKACA, PRKACA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5566 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17612 | |
| PRKACA | |
| PRKACA Fusion Protein Ag26392 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |