Learn More
PRKACA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24503-1-AP-150UL
Description
The PRKACA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PRKACA. This antibody recognizes human, mouse, rat antigen. The PRKACA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PRKACA | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 5566 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |