missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXI1 Monoclonal Antibody (OTI2C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800139S
This item is not returnable.
View return policy
Description
FOXI1 Monoclonal antibody specifically detects FOXI1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| FOXI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HC62 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 59343 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C1 | |
| Unconjugated | |
| SENP2 | |
| 5830401E05Rik; Fkh10; FKHL10; Forkhead; forkhead box I1; forkhead box protein I1; forkhead homolog 10; forkhead-like 10; forkhead-related activator 6; forkhead-related protein FKHL10; Forkhead-related transcription factor 6; Foxi1; FREAC6; FREAC-6; Hepatocyte nuclear factor 3 forkhead homolog 3; HFH3; HFH-3; HNF-3/forkhead homolog 3; HNF-3/fork-head homolog 3; HNF-3/fork-head homolog-3; MGC34197 | |
| Human recombinant protein fragment corresponding to amino acids 139-523 of human SENP2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction