missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKAR1B Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502750S
This item is not returnable.
View return policy
Description
PRKAR1B Monoclonal antibody specifically detects PRKAR1B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| PRKAR1B | |
| Monoclonal | |
| 0.57 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95182 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4701 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A3 | |
| Unconjugated | |
| Ndufa7 | |
| AI385716; cAMP dependent protein kinase, subunit type 1 beta; cAMP dependent regulatory type I beta subunit transcript 2; cAMP dependent regulatory type I beta subunit transcript 3; cAMP dependent regulatory type I beta subunit transcript 4; cAMP-dependent protein kinase type I-beta regulatory subunit; PRKAR1; PRKAR1B; protein kinase cAMP-dependent type I regulatory subunit beta; protein kinase, cAMP dependent regulatory, type I beta; protein kinase, cAMP dependent regulatory, type I, beta; protein kinase, cAMP-dependent, regulatory subunit type I beta; protein kinase, cAMP-dependent, regulatory, type I, beta; RGD1563094; RIbeta | |
| Full length human recombinant protein of human NDUFA7 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction