Learn More
STK38L Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI4D8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505176
Encompass_Preferred
Product Code. 17573312
  • € 540.87 / 100µL
Description
STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal. Northern blot analysis revealed expression of a 3.9-kb transcript in all tissues tested, with the possible exception of adult brain. Highest expression was in peripheral blood leukocytes. Immunoblot and immunofluorescence microscopy demonstrated predomitly nuclear expression of a 55-kD protein.
Specifications
Specifications
STK38L
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q7TSE6, Q9Y2H1
Stk38l
Human recombit protein fragment corresponding to amino acids 1-281 of human STK38L produced in E.coli.
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4D8
Unconjugated
STK38L
NDR2
Mouse
Affinity Chromatography
RUO
23012, 232533, 477663, 691337
-20° C, Avoid Freeze/Thaw Cycles