missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKCDBP Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81847IR
This item is not returnable.
View return policy
Description
PRKCDBP Polyclonal antibody specifically detects PRKCDBP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PRKCDBP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| Cavin-3, hSRBC, MGC20400, protein kinase C, delta binding protein, sdr-related gene product that binds to c-kinase, Serum deprivation response factor-related gene product that binds to C-kinase, SRBCprotein kinase C delta-binding protein | |
| Rabbit polyclonal PRKCDBP antibody was raised against an 17 amino acid peptide near the center of human PRKCDBP. The immunogen is located within amino acids 150 - 200 of PRKCDBP. Amino Acid Squence: ESSDEEPVESRAQRLRRT | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism | |
| 112464 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction