missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKD2 Monoclonal Antibody (OTI3A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501671
This item is not returnable.
View return policy
Description
Protein kinase D2 (PKD2) is a novel human serine threonine protein kinase gene. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKC and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.Specifications
| PRKD2 | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BZL6 | |
| PRKD2 | |
| Full length human recombit protein of human PRKD2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| OTI3A1 | |
| Unconjugated | |
| PRKD2 | |
| HSPC187, PKD2, nPKC-D2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 25865 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction