Learn More
Invitrogen™ PRKD2 Monoclonal Antibody (OTI4F1)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525231
Product Code. 15964834
  • € 588.00 / 100µL
Description
PRKD2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
PRKD2 (PKD2) is a member of the protein kinase D serine/threonine protein kinase family. PRKD2 is a component in the signal transduction pathway induced by phorbol esters and the CCK b gastrin receptor. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKC and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRKD2
Monoclonal
0.89 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BZL6
PRKD2
Full length human recombinant protein of PRKD2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI4F1
Unconjugated
PRKD2
AI325941; HSPC187; nPKC-D2; PKD2; Prkd2; protein kinase D2; RGD1308054; serine/threonine-protein kinase D2
Mouse
Affinity Chromatography
RUO
25865
-20°C, Avoid Freeze/Thaw Cycles