missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKD3/nPKC nu Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97269B
This item is not returnable.
View return policy
Description
PRKD3/nPKC nu Polyclonal antibody specifically detects PRKD3/nPKC nu in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PRKD3/nPKC nu | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Biotin | |
| EC 2.7.11, EC 2.7.11.13, EPK2PKD3, nPKC-nu, PKC-NU, PRKCNnPKC-NU, Protein kinase C nu type, protein kinase C, nu, protein kinase D3, Protein kinase EPK2, protein-serine/threonine kinase, serine/threonine-protein kinase D3 | |
| Produced in rabbits immunized with E. coli-derived Human PRKD3/nPKC nu fragment. | |
| 0.1 mL | |
| Cancer, Diabetes Research, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| 23683 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction