missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKD3/nPKC nu Antibody (OTI3D8), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73595PCP
This item is not returnable.
View return policy
Description
PRKD3/nPKC nu Monoclonal antibody specifically detects PRKD3/nPKC nu in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PRKD3/nPKC nu | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI3D8 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.13, EPK2PKD3, nPKC-nu, PKC-NU, PRKCNnPKC-NU, Protein kinase C nu type, protein kinase C, nu, protein kinase D3, Protein kinase EPK2, protein-serine/threonine kinase, serine/threonine-protein kinase D3 | |
| Human recombinant protein fragment corresponding to amino acids 300-508 of human PRKD3 (NP_005804) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Diabetes Research, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| 23683 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction