missing translation for 'onlineSavingsMsg'
Learn More

PRKG1 Monoclonal Antibody (OTI9A4), TrueMAB™, OriGene

Product Code. 17593591
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17593591 30 μL 30µL
17573591 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17593591 Supplier Origene Technologies Supplier No. TA501208S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRKG1 Monoclonal antibody specifically detects PRKG1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen PRKG1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI9A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pde4a
Gene Accession No. P27815
Gene Alias AAT8; AW125416; cGK; cGK 1; CGK 1 alpha; cGK 1 beta; cGK cGK cGKI; cGK cGK1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; cGMP dependent protein kinase type 1; cGMP kinase type I alpha; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase 1, alpha isozyme; cGMP-dependent protein kinase 1, beta isozyme; cGMP-dependent protein kinase I; Gm19690; OTTHUMP00000019618; PKG; Pkgi; Prkg1; PRKG1B; PRKGR1A; PRKGR1B; protein kinase cGMP-dependent 1; protein kinase, cGMP-dependent, regulatory, type I, beta; protein kinase, cGMP-dependent, type 1; protein kinase, cGMP-dependent, type I; RP11-346D6.1; Unknown (protein for MGC:134126); unnamed protein product
Host Species Mouse
Immunogen Full length human recombinant protein of human PDE4A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5141
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.