missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKG2 Monoclonal Antibody (OTI10D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807705
This item is not returnable.
View return policy
Description
cGKII is thought to play a key role in a diverse set of physiological pathway. cGKII may mediate intestinal secretion of water and electrolytes induced by the E. coli toxin STa and the intestinal peptide guanylin. Edentification of the pathway that mediates intestinal fluid secretion by E. coli STa has potential medical implications because STa causes traveler′s diarrhea and about 50% of infant mortality in developing countries. Transfection experiments in human cells disclose that cGKII phosphorylates SOX9 and attenuates SOX9 function by inhibiting its nuclear entry. Impaired differentiation of cultured KMI chondrocytes can be restored by silencing Sox9 by RNA interference. cGKII is postulated to be a molecular switch that couples the cessation of proliferation and the start of hypertrophic chondrocyte differentiation through attenuating SOX9 function.Specifications
| PRKG2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13237 | |
| PRKG2 | |
| Human recombit protein fragment corresponding to amino acids 40-320 of human PRKG2 (NP_006250 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI10D5 | |
| Unconjugated | |
| PRKG2 | |
| PRKGR2, cGK2, cGKII | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5593 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction