missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKG2 Monoclonal Antibody (OTI10D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807705S
This item is not returnable.
View return policy
Description
PRKG2 Monoclonal antibody specifically detects PRKG2 in Human samples. It is validated for Western BlotSpecifications
| PRKG2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P18846 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 466 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10D5 | |
| Unconjugated | |
| ATF1 | |
| AW212535; cGK 2; cGK2; cGKII; cGK-II; cGMP dependent protein kinase type II; cGMP-dependent protein kinase 2; cGMP-dependent protein kinase II; EC 2.7.11.12; GDPKII; PRKG2; PRKGR2; protein kinase cGMP- dependent type 2; protein kinase, cGMP- dependent, type 2; protein kinase, cGMP- dependent, type II; protein kinase, cGMP-dependent, type II; testicular tissue protein Li 99; type II cGMP-dependent protein kinase | |
| Full length human recombinant protein of human ATF1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction