missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT2 Mouse anti-Human, Clone: OTI3A3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503643
This item is not returnable.
View return policy
Description
PRMT is a member of the mammalian PRMT (protein arginine N-methyltransferase) family. The family consists of 7 members that share two large domains of homology. Outside of these domains, epitopes are identified and antibodies against all 7 PRMT members have been developed.These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins.Specifications
| PRMT2 | |
| Monoclonal | |
| 0.51 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P55345 | |
| PRMT2 | |
| Full length human recombit protein of human PRMT2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A3 | |
| Unconjugated | |
| PRMT2 | |
| HRMT1L1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3275 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction