Learn More
PRMT2 Mouse anti-Human, Unconjugated, Clone: 1F1B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66885-1-IG-150UL
This item is not returnable.
View return policy
Description
The PRMT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRMT2. This antibody recognizes human, mouse, rat antigen. The PRMT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PRMT2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot | |
| 1F1B9 | |
| Unconjugated | |
| 150 μL | |
| 3275 | |
| IgG2a |