missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRMT2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142908
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_001526.2; NP_001229793.1; NP_001229794.1; NP_001229795.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Arginine methyltransferase that methylates the guanidino nitrogens of arginyl residues in proteins such as STAT3, FBL, histone H4. Acts as a coactivator (with NCOA2) of the androgen receptor (AR)-mediated transactivation. Acts as a coactivator (with estrogen) of estrogen receptor (ER)-mediated transactivation. Enhances PGR, PPARG, RARA-mediated transactivation. May inhibit NF-kappa-B transcription and promote apoptosis. Represses E2F1 transcriptional activity (in a RB1-dependent manner). May be involved in growth regulation. [UniProt].
Specifications
| PRMT2 | |
| Polyclonal | |
| Unconjugated | |
| PRMT2 | |
| histone-arginine N-methyltransferase PRMT2; HMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 1; HMT1 hnRNP methyltransferase-like 1; Hrmt1l1; MGC111373; Prmt2; PRMT2 alpha; PRMT2 beta; PRMT2 gamma; protein arginine methyltransferase 2; protein arginine N-methyltransferase 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100294709, 15468, 3275, 499420 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P55345, Q9R144 | |
| PRMT2 | |
| Peptide with sequence C-DQPRTTKYHSVILQ. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction