missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 724.00
Specifications
| Antigen | PRMT3 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230617
|
Novus Biologicals
NBP3-35304-100ul |
100 μL |
€ 724.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229487
|
Novus Biologicals
NBP3-35304-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRMT3 Polyclonal antibody specifically detects PRMT3 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| PRMT3 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 10196 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human PRMT3 (NP_005779.1).,, Sequence:, MCSLASGATGGRGAVENEEDLPELSDSGDEAAWEDEDDADLPHGKQQTPCLFCNRLFTSAEETFSHCKSEHQFNIDSMVHKHGLEFYGYIKLINFIRLKNPTVEYMNSIYNPVPWEKEEYLKPVLEDDLLLQFDVEDLYEPVSVPFSYPNGLSENTSVVEKLKHMEARALSAEAALARAREDLQKMKQFAQDFVMHTDVRTCSSSTSVIADLQEDEDGVY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title