missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT3 Antibody (RAB-C372), DyLight 680 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-20143FR
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PRMT3 Monoclonal antibody specifically detects PRMT3 in Human samples. It is validated for ELISA
Specifica
| PRMT3 | |
| Monoclonal | |
| DyLight 680 | |
| EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| 10196 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA | |
| RAB-C372 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto